5-(4-Chlorophenyl)-2-furoic acid, 95+%, Thermo Scientific Chemicals
5-(4-Chlorophenyl)-2-furoic acid, 95+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-(4-Chlorophenyl)-2-furoic acid, 95+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 375540050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS41019-45-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only