You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Molecular Biology Reagents and Kits
Nucleic Acid Labeling and Detection
North2South™ Hybridization Stringency Wash Buffer (2X)
Thermo Scientific™
North2South™ Hybridization Stringency Wash Buffer (2X)
Catalog number:
37555
Related applications:
Nucleic Acid Gel Electrophoresis & Blotting
Have Questions?
Contact Us
Thermo Scientific™
North2South™ Hybridization Stringency Wash Buffer (2X)
Catalog number:
37555
Related applications:
Nucleic Acid Gel Electrophoresis & Blotting
Have Questions?
Contact Us
Catalog Number
37555
Unit Size
375 mL
Price (USD)
178.00
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Availability
Quantity
37555
375 mL
178.00
-
Save to list
Add to cart
Request bulk or custom format
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific North2South Hybridization Stringency Wash Buffer (2X), for use with the North2South Chemiluminescent Hybridization and Detection Kit.
Related Products
North2South™ Complete Biotin Random Prime Labeling and Detection Kit
North2South™ Chemiluminescent Hybridization and Detection Kit
North2South™ Chemiluminescent Substrate for HRP
North2South™ Hybridization Buffer
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer
Wash Buffer
Concentration
2X
Product Line
North2South™
Product Type
Wash Buffer
Quantity
375 mL
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search