Thermo Scientific™

Blocker™ FL Fluorescent Blocking Buffer (10X)

Catalog number: 37565
Have Questions?
Thermo Scientific™

Blocker™ FL Fluorescent Blocking Buffer (10X)

Catalog number: 37565
Have Questions?
Catalog Number
37565
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
37565100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Blocker FL Fluorescent Blocking Buffer is designed to block excess nonspecific binding sites to help reduce background fluorescence in western blotting applications. Blocker FL buffer helps enhance fluorescent protein detection by increasing the signal-to-noise ratio in blotting applications where background fluorescence may otherwise interfere with signal detection.

Compare and view all available blocking buffers

Working reagent is prepared by adding 1 part Blocker FL buffer to 9 parts ultrapure water. Optimal concentration should be determined empirically for each system. Compatible with both nitrocellulose and PVDF membranes.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Blocker FL
Quantity
100 mL
Form
Liquid
For Use With (Application)
Western Blot
Product Line
Blocker™
Concentration
10X
Shipping Condition
Wet Ice

Contents & Storage

Store in refrigerator (2°C to 8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text