Thermo Scientific Chemicals

2-Fluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 375780050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Fluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 375780050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 375780050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2905-21-7
IUPAC Name
2-fluorobenzene-1-sulfonyl chloride
Molecular Formula
C6H4ClFO2S
InChI Key
ZSZKAQCISWFDCQ-UHFFFAOYSA-N
SMILES
FC1=CC=CC=C1S(Cl)(=O)=O
Infrared spectrum
Conforms
Refractive index
1.5350 to 1.5390 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow-brown
Appearance (Form)
Liquid or solid
Titration Argentometric
>=96.0 % (ex Cl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text