Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | SuperBlock (TBS) Blocking Buffer, 100 mL |
Product Type | Blocking Buffer |
Quantity | 100 mL |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | SuperBlock™ |
Concentration | 1X |
Buffer | TBS |
Blocking Agent | Single purified protein |
Unit Size | 100 mL |
Description | SuperBlock (PBS) Blocking Buffer, 100 mL |
Product Type | Blocking Buffer |
Quantity | 100 mL |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | SuperBlock™ |
Concentration | 1X |
Buffer | PBS |
Blocking Agent | Single purified protein |
Unit Size | 100 mL |
Description | SuperBlock (PBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | SuperBlock™ |
Concentration | 1X |
Buffer | PBS |
Blocking Agent | Single purified protein |
Unit Size | 1 L |
Description | SuperBlock (PBS) Blocking Buffer, 5 L |
Product Type | Blocking Buffer |
Quantity | 5 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | SuperBlock™ |
Concentration | 1X |
Buffer | PBS |
Blocking Agent | Single purified protein |
Unit Size | 5 L |
Description | SuperBlock (TBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | SuperBlock™ |
Concentration | 1X |
Buffer | TBS |
Blocking Agent | Single purified protein |
Unit Size | 1 L |
Description | SuperBlock (TBS) Blocking Buffer, dry blend, 5 packs |
Product Type | Blocking Buffer, dry blend |
Quantity | 5 Pk. |
Form | Powder |
For Use With (Application) | ELISA, Western Blot |
Product Line | SuperBlock™ |
Buffer | TBS |
Blocking Agent | Single purified protein |
Unit Size | 5 packs |
Description | SuperBlock T20 (PBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | SuperBlock™ |
Concentration | 1X |
Buffer | PBST |
Blocking Agent | Single purified protein |
Unit Size | 1 L |
Description | SuperBlock T20 (TBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | SuperBlock™ |
Concentration | 1X |
Buffer | TBST |
Blocking Agent | Single purified protein |
Unit Size | 1 L |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
37581 | 100 mL | SuperBlock (TBS) Blocking Buffer, 100 mL | 52.65 Online price 56.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37580 | 100 mL | SuperBlock (PBS) Blocking Buffer, 100 mL | 52.65 Online price 56.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37515 | 1 L | SuperBlock (PBS) Blocking Buffer, 1 L | 226.65 Online price 245.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37518 | 5 L | SuperBlock (PBS) Blocking Buffer, 5 L | 735.65 Online price 794.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37535 | 1 L | SuperBlock (TBS) Blocking Buffer, 1 L | 232.65 Online price 251.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37545 | 5 packs | SuperBlock (TBS) Blocking Buffer, dry blend, 5 packs | 187.65 Online price 202.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37516 | 1 L | SuperBlock T20 (PBS) Blocking Buffer, 1 L | 250.65 Online price 270.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37536 | 1 L | SuperBlock T20 (TBS) Blocking Buffer, 1 L | 250.65 Online price 270.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|