Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Protein-Free (TBS) Blocking Buffer, 100 mL |
Product Type | Blocking Buffer, protein-free |
Quantity | 100 mL |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot, Arrays |
Concentration | 1X |
Buffer | TBS |
Blocking Agent | Non-protein compound |
Unit Size | 100 mL |
Description | Protein-Free (PBS) Blocking Buffer, 100 mL |
Product Type | Blocking Buffer, protein-free |
Quantity | 100 mL |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot, Arrays |
Concentration | 1X |
Buffer | PBS |
Blocking Agent | Non-protein compound |
Unit Size | 100 mL |
Description | Protein-Free (PBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer, protein-free |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot, Arrays |
Product Line | Pierce™ |
Concentration | 1X |
Buffer | PBS |
Blocking Agent | Non-protein compound |
Unit Size | 1 L |
Description | Protein-Free (TBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer, protein-free |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot, Arrays |
Product Line | Pierce™ |
Concentration | 1X |
Buffer | TBS |
Blocking Agent | Non-protein compound |
Unit Size | 1 L |
Description | Protein-Free T20 (PBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer, protein-free |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | Pierce™ |
Concentration | 1X |
Buffer | PBST |
Blocking Agent | Non-protein compound |
Unit Size | 1 L |
Description | Protein-Free T20 (TBS) Blocking Buffer, 1 L |
Product Type | Blocking Buffer, protein-free |
Quantity | 1 L |
Form | Liquid |
For Use With (Application) | ELISA, Western Blot |
Product Line | Pierce™ |
Concentration | 1X |
Buffer | TBST |
Blocking Agent | Non-protein compound |
Unit Size | 1 L |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
37585 | 100 mL | Protein-Free (TBS) Blocking Buffer, 100 mL | 51.65 Online price 55.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37584 | 100 mL | Protein-Free (PBS) Blocking Buffer, 100 mL | 51.65 Online price 55.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37572 | 1 L | Protein-Free (PBS) Blocking Buffer, 1 L | 223.65 Online price 244.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37570 | 1 L | Protein-Free (TBS) Blocking Buffer, 1 L | 227.65 Online price 234.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37573 | 1 L | Protein-Free T20 (PBS) Blocking Buffer, 1 L | 244.65 Online price 262.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37571 | 1 L | Protein-Free T20 (TBS) Blocking Buffer, 1 L | 251.65 Online price 265.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|