Thermo Scientific Chemicals

4-Nitrobenzyl chloroformate, 95%, Thermo Scientific Chemicals

Catalog number: 376040050
5 g, Each
Thermo Scientific Chemicals

4-Nitrobenzyl chloroformate, 95%, Thermo Scientific Chemicals

Catalog number: 376040050
5 g, Each
Quantity
Catalog number: 376040050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4457-32-3
IUPAC Name
(4-nitrophenyl)methyl carbonochloridate
Molecular Formula
C8H6ClNO4
InChI Key
MHSGOABISYIYKP-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(COC(Cl)=O)C=C1
Appearance (Form)
Solid and/or chunks or liquid after melting
Appearance (Color)
Off-white to yellow to beige
Infrared spectrum
Conforms
Melting point
30°C to 36°C
Titration Argentometric
>=94 % (ex Cl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text