Thermo Scientific Chemicals

Benzyloxyacetaldehyde, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 376070050
5 g, Each
Thermo Scientific Chemicals

Benzyloxyacetaldehyde, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 376070050
5 g, Each
Quantity
Catalog number: 376070050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
60656-87-3
IUPAC Name
2-(benzyloxy)acetaldehyde
Molecular Formula
C9H10O2
InChI Key
NFNOAHXEQXMCGT-UHFFFAOYSA-N
SMILES
O=CCOCC1=CC=CC=C1
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
GC
>=94 %
Appearance (Form)
Liquid
Stabilizer
0.5 % hydroquinone

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text