(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98%, Thermo Scientific Chemicals
(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 376190010
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
120.00 
Save 17.35 (14%)
-
Add to cart
Quantity:
1 mL
Request bulk or custom format
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
120.00 
Save 17.35 (14%)
Add to cart
(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98%, Thermo Scientific Chemicals
Catalog number376190010
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
120.00 
Save 17.35 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-(methoxymethyl)pyrrolidine
Molecular FormulaC6H13NO
InChI KeyCHPRFKYDQRKRRK-UHFFFAOYNA-N
SMILESCOCC1CCCN1
Molecular Weight (g/mol)115.18
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-32.5° to -36.5° (20°C, 589 nm) (c=2 in aq HCl)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only