(R)-N-BOC-Prolinal, 97+%, Thermo Scientific Chemicals
(R)-N-BOC-Prolinal, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-N-BOC-Prolinal, 97+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 376230010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametert-butyl 2-formylpyrrolidine-1-carboxylate
Molecular FormulaC10H17NO3
InChI KeyYDBPZCVWPFMBDH-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC1C=O
Molecular Weight (g/mol)199.25
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Colorless to yellow
Specific optical rotation+90° to +102° (20°C, 589 nm) (c=1, CHCl3)
Refractive index1.4600 to 1.4650 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only