Thermo Scientific Chemicals

L-aspartic acid dimethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 376660050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-aspartic acid dimethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 376660050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 376660050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
32213-95-9
IUPAC Name
1,4-dimethyl 2-aminobutanedioate hydrochloridyl
Molecular Formula
C6H11ClNO4
InChI Key
KUSLQZQCHPWKKH-UHFFFAOYNA-N
SMILES
[Cl].COC(=O)CC(N)C(=O)OC
Appearance (Color)
White to off-white
Titration Argentometric
>=97.5 %
Specific optical rotation
+13° to +17° (20°C, 589 nm) (c=1, CH3OH)
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text