L-threonine methyleester hydrochloride, +98%, Thermo Scientific Chemicals
L-threonine methyleester hydrochloride, +98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-threonine methyleester hydrochloride, +98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 376690250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS39994-75-7
IUPAC Namemethyl (2S,3R)-2-amino-3-hydroxybutanoate hydrochloride
Molecular FormulaC5H12ClNO3
InChI KeyOZSJLLVVZFTDEY-HJXLNUONSA-N
SMILESCl.COC(=O)[C@@H](N)[C@@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-9.5° to -10.5° (20°C, 589 nm) (c=1, Methanol, 20°C)
Appearance (Form)Powder
Infrared spectrumConforms
Titration Argentometric>=98.0 % (On dry substance)
Water=<1.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only