3-Phenyl-1-propyne, 97%, stabilized, Thermo Scientific Chemicals
3-Phenyl-1-propyne, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Phenyl-1-propyne, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 376800250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10147-11-2
IUPAC Name(prop-2-yn-1-yl)benzene
Molecular FormulaC9H8
InChI KeyNGKSKVYWPINGLI-UHFFFAOYSA-N
SMILESC#CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.5260 to 1.5300 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only