Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals
Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 376860050
Price (USD)/ Each
30.40
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
30.40
Add to cart
Ethyl isothiocyanate, 96%, Thermo Scientific Chemicals
Catalog number376860050
Price (USD)/ Each
30.40
-
Add to cart
Chemical Identifiers
CAS542-85-8
IUPAC Nameisothiocyanatoethane
Molecular FormulaC3H5NS
InChI KeyHBNYJWAFDZLWRS-UHFFFAOYSA-N
SMILESCCN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.5100 to 1.5190 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only