Thermo Scientific Chemicals

2-Amino-6-bromopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 376871000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Amino-6-bromopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 376871000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 376871000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19798-81-3
IUPAC Name
2-amino-6-bromopyridin-1-ium
Molecular Formula
C5H6BrN2
InChI Key
BKLJUYPLUWUEOQ-UHFFFAOYSA-O
SMILES
NC1=CC=CC(Br)=[NH+]1
Appearance (Color)
Light yellow to light brown
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
86°C to 90°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text