5-Bromo-2-methoxypyridine, 97%, Thermo Scientific Chemicals
5-Bromo-2-methoxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-methoxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 376880250
Price (USD)/ Each
194.00
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
194.00
Add to cart
5-Bromo-2-methoxypyridine, 97%, Thermo Scientific Chemicals
Catalog number376880250
Price (USD)/ Each
194.00
-
Add to cart
Chemical Identifiers
CAS13472-85-0
IUPAC Name5-bromo-2-methoxypyridine
Molecular FormulaC6H6BrNO
InChI KeyXADICJHFELMBGX-UHFFFAOYSA-N
SMILESCOC1=CC=C(Br)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5530 to 1.5570 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only