Thermo Scientific Chemicals

4-Bromo-1H-imidazole, 97%, Thermo Scientific Chemicals

Catalog number: 376900010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromo-1H-imidazole, 97%, Thermo Scientific Chemicals

Catalog number: 376900010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 376900010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2302-25-2
IUPAC Name
5-bromo-1H-imidazole
Molecular Formula
C3H3BrN2
InChI Key
FHZALEJIENDROK-UHFFFAOYSA-N
SMILES
BrC1=CN=CN1
Appearance (Color)
Colorless to beige
Infrared spectrum
Conforms
Appearance (Form)
Powder or flakes
HPLC
>=96.0 %
Melting point
126°C to 132°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text