4-Bromo-1H-imidazole, 97%, Thermo Scientific Chemicals
4-Bromo-1H-imidazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-1H-imidazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 376900050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2302-25-2
IUPAC Name5-bromo-1H-imidazole
Molecular FormulaC3H3BrN2
InChI KeyFHZALEJIENDROK-UHFFFAOYSA-N
SMILESBrC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or flakes
Appearance (Color)Colorless to beige
Infrared spectrumConforms
HPLC>=96.0 %
Melting point126°C to 132°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only