Palmitoleic acid, 99%
Palmitoleic acid, 99%
Thermo Scientific Chemicals

Palmitoleic acid, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 376912500
Price (USD)/ Each
73.65
Online exclusive
81.50 
Save 7.85 (10%)
-
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
73.65
Online exclusive
81.50 
Save 7.85 (10%)
Add to cart
Palmitoleic acid, 99%
Catalog number376912500
Price (USD)/ Each
73.65
Online exclusive
81.50 
Save 7.85 (10%)
-
Add to cart
Chemical Identifiers
CAS373-49-9
IUPAC Name(9Z)-hexadec-9-enoic acid
Molecular FormulaC16H30O2
InChI KeySECPZKHBENQXJG-FPLPWBNLSA-N
SMILESCCCCCC\C=C/CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4570 to 1.4590 (20°C, 589 nm)
Appearance (Form)Clear liquid
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only