Thermo Scientific Chemicals

2,3-Dichloropyrazine, 98%, Thermo Scientific Chemicals

Catalog number: 377041000
100 g, Each
Thermo Scientific Chemicals

2,3-Dichloropyrazine, 98%, Thermo Scientific Chemicals

Catalog number: 377041000
100 g, Each
Quantity
Catalog number: 377041000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4858-85-9
IUPAC Name
2,3-dichloropyrazine
Molecular Formula
C4H2Cl2N2
InChI Key
MLCNOCRGSBCAGH-UHFFFAOYSA-N
SMILES
ClC1=NC=CN=C1Cl
Appearance (Color)
Clear colorless to white to light yellow
Appearance (Form)
Solid or liquid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text