Methyl-beta-cyclodextrin, average Mw 1310
Methyl-beta-cyclodextrin, average Mw 1310
Methyl-beta-cyclodextrin, average Mw 1310
Methyl-beta-cyclodextrin, average Mw 1310
Thermo Scientific Chemicals

Methyl-beta-cyclodextrin, average Mw 1310

CAS: 128446-36-6 | C54H94O35 | 1303.311 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 377110050
Price (USD)
43.65
Special offer
Online exclusive
Ends: 26-Sep-2025
54.20
Save 10.55 (19%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 26-Sep-2025
54.20
Save 10.55 (19%)
Each
Add to cart
Chemical Identifiers
CAS128446-36-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Specific optical rotation+154.0° to +165.0° (20°C, 589 nm) (c=5, water)
Degree of substitution1.7 to 1.9 (NMR)
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Share catalog number, name or link