Thermo Scientific Chemicals

Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals

Catalog number: 377110250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl-beta-cyclodextrin, average Mw 1310, Thermo Scientific Chemicals

Catalog number: 377110250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 377110250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128446-36-6
Appearance (Form)
Powder
Specific optical rotation
+154.0° to +165.0° (20°C, 589 nm) (c=5, water)
Degree of substitution
1.7 to 1.9 (NMR)
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text