Thermo Scientific Chemicals

4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 377251000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 377251000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 377251000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
873-77-8
IUPAC Name
bromo(4-chlorophenyl)magnesium
Molecular Formula
C6H4BrClMg
InChI Key
CDEMHJCJMMOFMB-UHFFFAOYSA-M
SMILES
ClC1=CC=C([Mg]Br)C=C1
Appearance (Form)
Slightly hazy solution
Titration Active base
0.95 to 1.15 M
Appearance (Color)
Yellow to green to brown
Titration Active base
21.0 to 25.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text