Diethylzinc, 1.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Diethylzinc, 1.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylzinc, 1.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 377318000
Price (USD)/ Each
479.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
564.00 
Save 84.35 (15%)
-
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
479.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
564.00 
Save 84.35 (15%)
Add to cart
Diethylzinc, 1.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Catalog number377318000
Price (USD)/ Each
479.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
564.00 
Save 84.35 (15%)
-
Add to cart
Chemical Identifiers
CAS557-20-0, 108-88-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration Complexometric1.4 to 1.6 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only