This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 377348000
Price (USD)/ Each
272.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
320.00
Save 47.35 (15%)
-
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
272.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
320.00
Save 47.35 (15%)
Add to cart
Cyclohexylmagnesium chloride, 1.3M solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Solution
Titration Grignard1.26 to 1.40 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.