Thermo Scientific Chemicals

Quinaldine red, pure

Catalog number: 377511000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Quinaldine red, pure

Catalog number: 377511000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 377511000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
117-92-0
IUPAC Name
2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide
Molecular Formula
C21H23IN2
InChI Key
JOLANDVPGMEGLK-UHFFFAOYSA-M
SMILES
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C
Appearance (Color)
Grey to green-black to dark blue
Appearance (Form)
Powder
UV spectrum
Complies
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text