AppearanceMay develop some turbidity or precipitate
Appearance (Form)Solution
Titration Argentometric0.85 to 1.00 M
Appearance (Color)Clear to slightly hazy colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.