Thermo Scientific Chemicals

Barium hydroxide octahydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 377820050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Barium hydroxide octahydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 377820050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 377820050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12230-71-6
IUPAC Name
barium(2+) octahydrate dihydroxide
Molecular Formula
BaH18O10
InChI Key
ZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
Appearance (Form)
Crystalline powder or crystals or flakes and/or
Appearance (Form)
chunks
Total trace metal impurities
=<0.05 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text