Sodium chromate, 98+%, anhydrous, Thermo Scientific Chemicals
Sodium chromate, 98+%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium chromate, 98+%, anhydrous, Thermo Scientific Chemicals

Sodium chromate, 98+%, CrNa2O4, CAS Number-7775-11-3, asex, atlacide, chlorate de sodium, chloric acid, sodium salt, chlorsaure, desolet, kusatol, rasikal, sodium chlorate, tumbleleaf, 500g, 98% min. (Iodometry), CHEBI:65242, Yellow, 231-889-5, 161.97, GHS Signal Word: Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 377875000
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
Add to cart
Sodium chromate, 98+%, anhydrous, Thermo Scientific Chemicals
Catalog number377875000
Price (USD)/ Each
150.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
177.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium dioxochromiumbis(olate)
Molecular FormulaCrNa2O4
InChI KeyPXLIDIMHPNPGMH-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][Cr]([O-])(=O)=O
Molecular Weight (g/mol)161.97
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration Iodometric>=98.0 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only