Thermo Scientific Chemicals

Hydridotetrakis(triphenylphosphine)rhodium(I), Thermo Scientific Chemicals

Catalog number: 377932500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

Hydridotetrakis(triphenylphosphine)rhodium(I), Thermo Scientific Chemicals

Catalog number: 377932500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 377932500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18284-36-1
IUPAC Name
tetrakis(triphenylphosphane) λ¹-rhodium
Molecular Formula
C72H61P4Rh
InChI Key
QTTJMGBPWXLZKV-UHFFFAOYSA-N
SMILES
[RhH].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Ochre
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Rhodium (Rh)
>=8.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text