Chemical Identifiers
CAS38465-86-0
IUPAC Nameiridium(1+) (1Z,5Z)-cycloocta-1,5-diene hexafluoro-λ⁵-phosphanuide bis(methyldiphenylphosphane)
Molecular FormulaC34H38F6IrP3
InChI KeyLXKHQEQLBSJJCO-JXNOXZOESA-N
SMILES[Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1
View more