Chemical Identifiers
CAS12354-84-6
IUPAC Namediiridium(3+) bis(pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride
Molecular FormulaC20H30Cl4Ir2
InChI KeyMMAGMBCAIFVRGJ-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3].C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C
View more