Thermo Scientific Chemicals

(Pentamethylcyclopentadienyl)iridium(III) Chloride dimer, 99%, Thermo Scientific Chemicals

Catalog number: 377952500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

(Pentamethylcyclopentadienyl)iridium(III) Chloride dimer, 99%, Thermo Scientific Chemicals

Catalog number: 377952500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 377952500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12354-84-6
IUPAC Name
diiridium(3+) bis(pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride
Molecular Formula
C20H30Cl4Ir2
InChI Key
MMAGMBCAIFVRGJ-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3].C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C
Appearance (Color)
Orange
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Assay
>=98.5 % (ex Ir)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text