Chemical Identifiers
CAS40691-33-6
IUPAC Namedichloropalladiumbis(ylium); bis(tris(2-methylphenyl)phosphane)
Molecular FormulaC42H42Cl2P2Pd
InChI KeyGMLMZLGOMMIIMA-UHFFFAOYSA-L
SMILESCl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
View more