Bis[tri(o-tolyl)phosphine]palladium(II) chloride, 95%, Thermo Scientific Chemicals
Bis[tri(o-tolyl)phosphine]palladium(II) chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis[tri(o-tolyl)phosphine]palladium(II) chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 377960050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS40691-33-6
IUPAC Namedichloropalladiumbis(ylium); bis(tris(2-methylphenyl)phosphane)
Molecular FormulaC42H42Cl2P2Pd
InChI KeyGMLMZLGOMMIIMA-UHFFFAOYSA-L
SMILESCl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Elemental Analysis>=94 % (ex C)
Appearance (Color)Yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only