Chemical Identifiers
CAS161265-03-8
IUPAC Name[5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphane
Molecular FormulaC39H32OP2
InChI KeyCXNIUSPIQKWYAI-UHFFFAOYSA-N
SMILESCC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
View more