Boron tribromide, 1M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Boron tribromide, 1M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron tribromide, 1M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 378191000
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
Add to cart
Boron tribromide, 1M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Catalog number378191000
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10294-33-4, 142-82-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Strength0.9 to 1.1 M
Appearance (Color)Clear yellow to amber
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only