Chemical Identifiers
CAS258278-25-0
IUPAC Name1,3-bis[2,6-bis(propan-2-yl)phenyl]-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium chloride
Molecular FormulaC27H39ClN2
InChI KeyLWPXTYZKAWSRIP-UHFFFAOYSA-M
SMILES[Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C
View more