Thermo Scientific Chemicals

Nickel(II) chloride, 99.99%, (trace metal basis), anhydrous, powder, Thermo Scientific Chemicals

Catalog number: 378350250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Nickel(II) chloride, 99.99%, (trace metal basis), anhydrous, powder, Thermo Scientific Chemicals

Catalog number: 378350250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 378350250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7718-54-9
IUPAC Name
nickel(2+) dichloride
Molecular Formula
Cl2Ni
InChI Key
QMMRZOWCJAIUJA-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ni++]
Appearance (Form)
Powder
Titration Complexometric
44.6 to 46.0 % (Ni)
Total trace metal impurities
=<100 ppm
Appearance (Color)
Yellow to orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text