Thermo Scientific Chemicals

Tetrapropylammonium perruthenate, Thermo Scientific Chemicals

Catalog number: 378390010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrapropylammonium perruthenate, Thermo Scientific Chemicals

Catalog number: 378390010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 378390010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
114615-82-6
IUPAC Name
tetrapropylazanium tetrahydrate ruthenium
Molecular Formula
C12H36NO4Ru
InChI Key
HUCLFLGLPCVDMZ-UHFFFAOYSA-N
SMILES
O.O.O.O.[Ru].CCC[N+](CCC)(CCC)CCC
Appearance (Color)
Black
Ruthenium (Ru)
>=27.7 %
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text