(R)-(-)-2-Phenylbutyric acid, 99%, Thermo Scientific Chemicals
(R)-(-)-2-Phenylbutyric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-2-Phenylbutyric acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 378520050
Price (USD)/ Each
609.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
609.00
Add to cart
(R)-(-)-2-Phenylbutyric acid, 99%, Thermo Scientific Chemicals
Catalog number378520050
Price (USD)/ Each
609.00
-
Add to cart
Chemical Identifiers
CAS938-79-4
IUPAC Name2-phenylbutanoic acid
Molecular FormulaC10H12O2
InChI KeyOFJWFSNDPCAWDK-UHFFFAOYNA-N
SMILESCCC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.5150 to 1.5170 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only