Thermo Scientific Chemicals

Iodomethylcyclopentane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 378640010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Iodomethylcyclopentane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 378640010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 378640010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27935-87-1
IUPAC Name
(iodomethyl)cyclopentane
Molecular Formula
C6H11I
InChI Key
DUMSKQUKLVSSII-UHFFFAOYSA-N
SMILES
ICC1CCCC1
Appearance (Color)
Colorless to yellow to light orange
Appearance (Form)
Liquid
Stabilizer
copper powder
Infrared spectrum
Conforms
Refractive index
1.5360 to 1.5380 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text