Iodomethylcyclopentane, 98%, stabilized, Thermo Scientific Chemicals
Iodomethylcyclopentane, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodomethylcyclopentane, 98%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 378640050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(iodomethyl)cyclopentane
Molecular FormulaC6H11I
InChI KeyDUMSKQUKLVSSII-UHFFFAOYSA-N
SMILESICC1CCCC1
Molecular Weight (g/mol)210.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to light orange
Appearance (Form)Liquid
Stabilizercopper powder
Infrared spectrumConforms
Refractive index1.5360 to 1.5380 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only