1x1 image pixel for data collection
N,N-Dimethylacetamide, 99.5%, extra pure
N,N-Dimethylacetamide, 99.5%, extra pure
N,N-Dimethylacetamide, 99.5%, extra pure
N,N-Dimethylacetamide, 99.5%, extra pure
Thermo Scientific Chemicals

N,N-Dimethylacetamide, 99.5%, extra pure

CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number 378710200
Price (USD)
831.65
Online Exclusive
924.00
Save 92.35 (10%)
Each
In stock
Add to cart
Quantity:
20 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)
831.65
Online Exclusive
924.00
Save 92.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS127-19-5
IUPAC NameN,N-dimethylacetamide
Molecular FormulaC4H9NO
InChI KeyFXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILESCN(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Free acid (CH3COOH)=<0.1 %
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
GC>=99.4 %
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection