Thermo Scientific Chemicals

tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals

Catalog number: 378720010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals

Catalog number: 378720010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 378720010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1634-04-4
IUPAC Name
2-methoxy-2-methylpropane
Molecular Formula
C5H12O
InChI Key
BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES
COC(C)(C)C
Refractive index
1.3675 to 1.3695 (20°C, 589 nm)
Water
=<0.03 %
Residue after evaporation
=<1 mg/100 ml
Impurity
=<0.10 % Total other organics
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text