tert-Butyl methyl ether, 99.9%, extra pure
tert-Butyl methyl ether, 99.9%, extra pure
tert-Butyl methyl ether, 99.9%, extra pure
tert-Butyl methyl ether, 99.9%, extra pure
Thermo Scientific Chemicals

tert-Butyl methyl ether, 99.9%, extra pure

CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number 378720100
Price (EUR)
191,25
Online Exclusive
225,00
Save 33,75 (15%)
Each
-
Add to cart
Quantity:
10 L
Packaging:
Plastic Drum
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
191,25
Online Exclusive
225,00
Save 33,75 (15%)
Each
Add to cart
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3675 to 1.3695 (20°C, 589 nm)
Water=<0.03 %
Residue after evaporation=<1 mg/100 ml
Impurity=<0.10 % Total other organics
Appearance (Form)Clear liquid
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0466034Certificate of AnalysisSep 10, 2024378720100, 378720025, 378720010
A0457574Certificate of AnalysisNov 27, 2023378720100, 378720025, 378720010
A0457575Certificate of AnalysisNov 17, 2023378720100, 378720025, 378720010
A0457573Certificate of AnalysisNov 14, 2023378720100, 378720025, 378720010
A0454726Certificate of AnalysisAug 25, 2023378720100, 378720025, 378720010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 2
Skin Corrosion/Irritation
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H315 - Causes skin irritation

Physical Hazards

H225 - Highly flammable liquid and vapor

Precautionary Statements

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P240 - Ground and bond container and receiving equipment

Share catalog number, name or link

1x1 image pixel for data collection