Benzo[b]furan-2-boronic acid, 97%, Thermo Scientific Chemicals
Benzo[b]furan-2-boronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzo[b]furan-2-boronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 378730010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98437-24-2
IUPAC Name(1-benzofuran-2-yl)boronic acid
Molecular FormulaC8H7BO3
InChI KeyPKRRNTJIHGOMRC-UHFFFAOYSA-N
SMILESOB(O)C1=CC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only