2-Ethylbenzoic acid, 97%, Thermo Scientific Chemicals
2-Ethylbenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethylbenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 378750010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS612-19-1
IUPAC Name2-ethylbenzoic acid
Molecular FormulaC9H10O2
InChI KeyCGMMPMYKMDITEA-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
NMR (1H-NMR)Consistent with structure
Melting point62°C to 66°C
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only