Thermo Scientific Chemicals

(Toluene)tricarbonylchromium, 98%, Thermo Scientific Chemicals

Catalog number: 378820050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(Toluene)tricarbonylchromium, 98%, Thermo Scientific Chemicals

Catalog number: 378820050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 378820050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12083-24-8
IUPAC Name
tris(methanidylidyneoxidanium) toluene chromium
Molecular Formula
C10H8CrO3
InChI Key
KEXFCQUTYPQKCG-UHFFFAOYSA-N
SMILES
[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].CC1=CC=CC=C1
Appearance (Color)
Yellow to light orange
Infrared spectrum
Conforms
Elemental Analysis
>=97.5 %
Melting point
78°C to 82°C
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text