Thermo Scientific Chemicals

Ethynylferrocene, 97%, Thermo Scientific Chemicals

Catalog number: 378830010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethynylferrocene, 97%, Thermo Scientific Chemicals

Catalog number: 378830010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 378830010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1271-47-2
Molecular Formula
C12H10Fe
SMILES
[Fe].c1cccc1.C#Cc1cccc1
Molecular Weight (g/mol)
210.06
MDL Number
MFCD03701501
HPLC
>=96.0 %
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Appearance (Color)
Red-brown
Melting point
50°C to 57°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text